siRNA Details
Gene Symbol: Lcn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276448.1 | NP_001263377.1 | 5, 6 | 551 | ||
GenBank mRNA | AK033743.1 | - | 3, 4 | 1079 | ||
FJ465144.1 | - | 5, 6 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
