siRNA Details
Gene Symbol: Dido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291433.1 | NP_001278362.1 | 7 | 2177 | ||
NM_175551.4 | NP_780760.2 | 7 | 2177 | |||
NM_177852.4 | NP_808520.2 | 7 | 2125 | |||
GenBank mRNA | AK049707.1 | - | 1 | 792 | ||
AK129117.1 | - | 7 | 2125 | |||
AY425951.1 | - | 7 | 2079 | |||
AY425952.1 | - | 7 | 2079 | |||
BC044755.1 | - | 1 | 276 | |||
BC138712.1 | - | 6 | 1954 | 8861734 | ||
BC138713.1 | - | 6 | 1954 | 8861735 | ||
BC145401.1 | - | 6 | 1955 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
