siRNA Details
Gene Symbol: Sdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177879.5 | NP_808547.3 | 4 | 954 | ||
GenBank mRNA | AK030635.1 | - | 4 | 465 | ||
AK147578.1 | - | 4 | 999 | |||
BC060237.1 | - | 4 | 538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
