siRNA Details
Gene Symbol: A530040E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142678.1 | - | 358 |
Gene Symbol: Dcdc2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177964.2 | NP_001171435.2 | 9 | 850 | ||
GenBank mRNA | AK003729.1 | - | 9 | 851 |
Gene Symbol: F830002E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089566.1 | - | 1 | 1932 | ||
AK172269.1 | - | 1 | 1932 |
Gene Symbol: Gm11525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080724.1 | - | 1 | 232 |
Gene Symbol: Gm16938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085722.1 | - | 408 | |||
BC144932.1 | - | 4 | 496 | |||
BC144933.1 | - | 4 | 469 | |||
BC144936.1 | - | 4 | 472 |
Gene Symbol: Gm38947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133700.1 | - | 3 | 1925 |
Gene Symbol: Gm7102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177513.1 | NP_001170984.1 | 1 | 1139 | ||
GenBank mRNA | AK163227.1 | - | 1 | 1143 |
Gene Symbol: Rpia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK166785.1 | - | 2 | 1609 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK145457.1 | - | 3591 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
