siRNA Details
Gene Symbol: 4933402J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177901.3 | NP_808569.1 | 2 | 300 | ||
GenBank mRNA | AK077084.1 | - | 2 | 301 | ||
AK132946.1 | - | 2 | 355 | |||
BC117078.1 | - | 2 | 293 | 40126397 | ||
BC120779.1 | - | 2 | 293 | 40129702 |
