siRNA Details
Gene Symbol: Gsdmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183194.3 | NP_899017.2 | 4 | 425 | ||
GenBank mRNA | AK037079.1 | - | 4 | 426 |
Gene Symbol: Gsdmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168274.1 | NP_001161746.1 | 609 | |||
NM_177912.4 | NP_808580.2 | 374 | ||||
GenBank mRNA | AK033603.1 | - | 375 | |||
BC113153.1 | - | 367 | 40058717 | |||
BC113155.1 | - | 595 | 40058514 |
Gene Symbol: Gsdmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028992.1 | NP_083268.1 | 3 | 374 | ||
GenBank mRNA | AK018537.1 | - | 3 | 375 | ||
AK163061.1 | - | 3 | 375 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC099685.1 | - | 469 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
