siRNA Details
Gene Symbol: BC080696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC080696.1 | - | 1 | 271 |
Gene Symbol: A430089I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177913.4 | NP_808581.1 | 2 | 346 | ||
GenBank mRNA | AK040373.1 | - | 2 | 347 | ||
BC147369.1 | - | 2 | 313 | 9056433 |
Gene Symbol: BC061212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198667.1 | NP_941069.1 | 2 | 346 | ||
GenBank mRNA | BC061212.1 | - | 2 | 346 | 30250942 |
Gene Symbol: Gm7978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270457.1 | NP_001257386.1 | 2 | 346 |
Gene Symbol: LOC545763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN677185.1 | - | 1 | 240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
