siRNA Details
Gene Symbol: Igsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177591.4 | NP_808259.2 | 4 | 464 | ||
NM_177915.4 | NP_808583.1 | 4 | 464 | |||
NM_183335.2 | NP_899178.2 | 3 | 328 | |||
GenBank mRNA | AK030452.1 | - | 4 | 436 | ||
AK129126.1 | - | 4 | 446 | |||
AY227771.1 | - | 4 | 458 | |||
AY227772.1 | - | 4 | 458 | |||
AY227773.1 | - | 3 | 328 | |||
BC089474.1 | - | 4 | 445 | |||
BC100420.1 | - | 4 | 464 |

More Information
Other Species Targeted

Gene Ontology Categories:
