siRNA Details
Gene Symbol: EDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005609.1 | NP_001005609.1 | 8 | 3314 | ||
NM_001005612.2 | NP_001005612.2 | 8 | 3305 | |||
NM_001399.4 | NP_001390.1 | 8 | 3320 | |||
XM_006724630.2 | XP_006724693.1 | 8 | 3292 | |||
XM_017029336.1 | XP_016884825.1 | 8 | 3259 | |||
GenBank mRNA | AF040628.1 | - | 8 | 3321 | ||
AF061189.1 | - | 8 | 3315 | |||
AK311532.1 | - | 1 | 933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
