siRNA Details
Gene Symbol: Gm16938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085722.1 | - | 821 | |||
BC030866.1 | - | 1 | 58 | |||
BC040816.1 | - | 1 | 46 | |||
BC144932.1 | - | 4 | 909 | |||
BC144933.1 | - | 4 | 882 | |||
BC144936.1 | - | 4 | 885 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
