siRNA Details
Gene Symbol: Dgkk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177914.3 | NP_808582.3 | 6614 | |||
GenBank mRNA | AK081332.1 | - | 1 | 1822 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
