siRNA Details
Gene Symbol: Setd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178029.3 | NP_821172.2 | 9, 10 | 2875 | ||
GenBank mRNA | BC049883.1 | - | 2, 3 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
