siRNA Details
Gene Symbol: Rassf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178045.4 | NP_835146.3 | 10 | 1029 | ||
GenBank mRNA | AK036528.1 | - | 10 | 993 | ||
AK089460.1 | - | 10 | 987 | |||
AK139284.1 | - | 10 | 976 | |||
AK151561.1 | - | 10 | 994 | |||
AK152639.1 | - | 10 | 994 | |||
AK160584.1 | - | 10 | 1003 | |||
AK164357.1 | - | 10 | 992 | |||
AK170955.1 | - | 10 | 978 | |||
AK171198.1 | - | 10 | 1006 |

More Information
Other Species Targeted

Gene Ontology Categories:
