siRNA Details
Gene Symbol: Etl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081006.1 | NP_001074475.1 | 7 | 1459 | ||
NM_001177630.2 | NP_001171101.2 | 6 | 1557 | |||
NM_001177631.1 | NP_001171102.1 | 6 | 1557 | |||
NM_029895.4 | NP_084171.2 | 7 | 1770 | |||
NM_178059.5 | NP_835160.2 | 7 | 1770 | |||
GenBank mRNA | AB093289.2 | - | 6 | 1347 | ||
AB125594.2 | - | 7 | 1770 | |||
AB125595.2 | - | 7 | 1770 | |||
AK052020.1 | - | 8 | 1872 | |||
AK054453.1 | - | 2 | 713 | |||
AK086078.1 | - | 2 | 714 | |||
AK147510.1 | - | 6 | 1464 | |||
AK158471.1 | - | 2 | 505 | |||
BC158051.1 | - | 6 | 1279 | 9007405 | ||
BC172143.1 | - | 6 | 1277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
