siRNA Details
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178080.4 | NP_835181.2 | 1 | 633 | ||
GenBank mRNA | AK028810.1 | - | 1 | 601 | ||
AK042081.1 | - | 1 | 609 | |||
AK122286.1 | - | 1 | 628 | |||
BC025653.1 | - | 1 | 559 | |||
BC050783.1 | - | 1 | 587 | |||
BC057350.1 | - | 1 | 550 | |||
BC141172.1 | - | 1 | 561 |
