siRNA Details
Gene Symbol: Smg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178246.3 | NP_839977.2 | 13, 14 | 2206 | ||
GenBank mRNA | AK129286.1 | - | 13, 14 | 2185 | ||
AK133688.1 | - | 4, 5 | 1030 | |||
AK143911.1 | - | 13, 14 | 2208 | |||
AK172217.1 | - | 12, 13 | 2086 | |||
BC048378.1 | - | 2, 3 | 386 | |||
BC066787.1 | - | 8, 9 | 1576 |

More Information
Other Species Targeted

Gene Ontology Categories:
