siRNA Details
Gene Symbol: Gm13102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085419.1 | NP_001078888.1 | 2 | 255 | ||
GenBank mRNA | BC132592.1 | - | 2 | 256 | 40130869 | |
BC132620.1 | - | 2 | 256 | 40130897 |
Gene Symbol: Gm13083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126324.1 | NP_001119796.1 | 1 | 188 |
Gene Symbol: Pramef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085414.2 | NP_001078883.2 | 2 | 208 |
Gene Symbol: Pramel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001319.3 | NP_001001319.2 | 2 | 256 | ||
GenBank mRNA | AF490340.1 | - | 1 | 188 | ||
AK145585.1 | - | 2 | 257 | |||
AK163305.1 | - | 2 | 257 |
Gene Symbol: Pramel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085418.2 | NP_001078887.1 | 2 | 239 | ||
GenBank mRNA | BC052827.1 | - | 2 | 249 | 30033377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
