siRNA Details
Gene Symbol: Abca13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178259.3 | NP_839990.2 | 20 | 8792 | ||
GenBank mRNA | AY160971.1 | - | 2 | 111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
