siRNA Details
Gene Symbol: Arap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178407.3 | NP_848494.2 | 33 | 6002 | ||
GenBank mRNA | AK084988.1 | - | 2 | 1523 | ||
AK122316.1 | - | 21 | 3302 | |||
AK134702.1 | - | 1 | 404 | |||
AK136522.1 | - | 6 | 1197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
