siRNA Details
Gene Symbol: Thnsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033929.2 | NP_001029101.1 | 9 | 1684 | ||
NM_178413.5 | NP_848500.3 | 10 | 1752 | |||
GenBank mRNA | AK165463.1 | - | 9 | 1650 | ||
BC025604.1 | - | 10 | 1715 | 5250319 | ||
BC051244.1 | - | 9 | 1646 | 4975661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
