siRNA Details
Gene Symbol: Nop16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178605.4 | NP_848720.1 | 5 | 883 | ||
GenBank mRNA | AK002696.1 | - | 5 | 831 | ||
AK003769.1 | - | 5 | 866 | |||
AK013251.1 | - | 5 | 884 | |||
AK088248.1 | - | 5 | 884 | |||
BC012213.1 | - | 5 | 859 | 3964770 |
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039200.2 | NP_001034289.1 | 23 | 3829 | ||
NM_146157.4 | NP_666269.2 | 24 | 3838 | |||
GenBank mRNA | AK049061.1 | - | 24 | 3831 | ||
AK077496.1 | - | 24 | 3829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
