siRNA Details
Gene Symbol: Prss33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081399.2 | NP_001074868.2 | 5 | 659 | ||
GenBank mRNA | AY262280.1 | - | 5 | 635 | ||
BC101950.1 | - | 5 | 635 | 40044230 | ||
BC103541.1 | - | 6 | 731 | 40044224 | ||
BC103542.1 | - | 5 | 635 | 40044219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
