siRNA Details
Gene Symbol: Pik3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF528078.1 | - | 5 | 824 | ||
AK028419.1 | - | 1 | 2913 | |||
AK029304.1 | - | 1 | 2902 | |||
AK160300.1 | - | 3 | 1895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
