siRNA Details
Gene Symbol: Smg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178246.3 | NP_839977.2 | 15 | 2303 | ||
GenBank mRNA | AK129286.1 | - | 15 | 2282 | ||
AK133688.1 | - | 6 | 1127 | |||
AK143911.1 | - | 15 | 2305 | |||
AK172217.1 | - | 14 | 2183 | |||
BC048378.1 | - | 4 | 483 | |||
BC066787.1 | - | 10 | 1673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
