siRNA Details
Gene Symbol: Gm13102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085419.1 | NP_001078888.1 | 3 | 771 | ||
GenBank mRNA | BC132592.1 | - | 3 | 772 | 40130869 | |
BC132620.1 | - | 3 | 772 | 40130897 |
Gene Symbol: Pramel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001319.3 | NP_001001319.2 | 3 | 772 | ||
GenBank mRNA | AF490340.1 | - | 3 | 656 | ||
AK145585.1 | - | 3 | 773 | |||
AK163305.1 | - | 3 | 631 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
