siRNA Details
Gene Symbol: Yy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098723.1 | NP_001092193.1 | 1 | 2149 | ||
GenBank mRNA | AK161451.1 | - | 1 | 2151 |
Gene Symbol: Mbtps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036071.1 | - | 6 | 3001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
