siRNA Details
Gene Symbol: Nrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178591.2 | NP_848706.2 | 2 | 728 | ||
GenBank mRNA | AK048708.1 | - | 2 | 910 | ||
AK051824.1 | - | 6 | 946 | |||
AK141037.1 | - | 2 | 916 | |||
AK170314.1 | - | 3 | 251 | |||
AY227027.1 | - | 1 | 62 | |||
AY648975.1 | - | 2 | 728 | |||
AY648976.1 | - | 6 | 563 | |||
BC151113.1 | - | 2 | 812 | 9088026 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001271122, NM_001271121, NM_031588, NM_001271119, NM_001271124, and others.
Gene Ontology Categories:
