siRNA Details
Gene Symbol: Ints7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034856.1 | - | 1, 2 | 197 | ||
AK085362.1 | - | 1, 2 | 234 | |||
AK086462.1 | - | 1, 2 | 227 | |||
CD348032.1 | - | 1, 2 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
