siRNA Details
Gene Symbol: Lrrc8d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122768.1 | NP_001116240.1 | 3 | 3215 | ||
NM_178701.3 | NP_848816.3 | 3 | 3205 | |||
GenBank mRNA | AK046249.1 | - | 3 | 3214 | ||
AK050400.1 | - | 3 | 3175 | |||
AK080722.1 | - | 3 | 3149 | |||
AK137459.1 | - | 3 | 3216 | |||
AK167985.1 | - | 2 | 2923 | |||
AK171882.1 | - | 1 | 115 | |||
AK207105.1 | - | 1 | 178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
