siRNA Details
Gene Symbol: Gm2042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270792.1 | NP_001257721.1 | 3 | 570 | ||
GenBank mRNA | AK135749.1 | - | 3 | 548 | ||
AK139641.1 | - | 3 | 547 | |||
BC086484.1 | - | 3 | 543 | 30476881 |
Gene Symbol: Gm10436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105254.1 | NP_001098724.1 | 3 | 622 | ||
GenBank mRNA | AK139576.1 | - | 2 | 207 | ||
BC094894.1 | - | 2 | 184 |
Gene Symbol: Oog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178657.5 | NP_848772.3 | 3 | 570 | ||
GenBank mRNA | AB050008.3 | - | 1 | 160 | ||
AB077322.2 | - | 1 | 160 | |||
AK145346.1 | - | 3 | 550 | |||
CF915726.1 | - | 3 | 570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
