siRNA Details
Gene Symbol: Pde12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178668.3 | NP_848783.3 | 1 | 1163 | ||
GenBank mRNA | AK051991.1 | - | 379 | |||
AK082947.1 | - | 1 | 986 | |||
AK088843.1 | - | 1 | 976 | |||
AK143567.1 | - | 1 | 990 | |||
AK155088.1 | - | 997 | ||||
AK161083.1 | - | 1 | 1203 | |||
AK167708.1 | - | 1 | 961 | |||
BC064450.1 | - | 1 | 1163 | 6417852 |
