siRNA Details
Gene Symbol: C130074G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178692.3 | NP_848807.1 | 2 | 371 | ||
GenBank mRNA | AK052654.1 | - | 2 | 335 | ||
AK081755.1 | - | 2 | 350 | |||
AK084264.1 | - | 2 | 344 | |||
AK087640.1 | - | 2 | 362 | |||
BC116951.1 | - | 2 | 371 | 40126270 | ||
BC116953.1 | - | 2 | 371 | 40126272 | ||
BY710149.1 | - | 2 | 553 |

More Information
Other Species Targeted

Gene Ontology Categories:
