siRNA Details
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178709.4 | NP_848824.1 | 3 | 799 | ||
GenBank mRNA | AK028768.1 | - | 3 | 434 | ||
AK030208.1 | - | 3 | 431 | |||
AK033867.1 | - | 3 | 295 | |||
AK046190.1 | - | 3 | 784 | |||
AK046997.1 | - | 424 | ||||
AK051527.1 | - | 3 | 800 | |||
BC094247.1 | - | 2 | 284 | 5717984 |
