siRNA Details
Gene Symbol: Samm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178614.4 | NP_848729.1 | 3 | 316 | ||
GenBank mRNA | AK003990.1 | - | 3 | 272 | ||
AK039949.1 | - | 3 | 316 | |||
AK041783.1 | - | 310 | ||||
AK084395.1 | - | 3 | 310 | |||
AK086633.1 | - | 308 | ||||
AK144975.1 | - | 3 | 317 | |||
AK159740.1 | - | 3 | 328 | |||
AK161378.1 | - | 3 | 306 | |||
AK167806.1 | - | 3 | 310 | |||
AK168510.1 | - | 3 | 272 | |||
AK169066.1 | - | 3 | 306 | |||
AK169351.1 | - | 3 | 273 | |||
BC119178.1 | - | 3 | 310 | 8733927 | ||
BC119180.1 | - | 3 | 310 | 8733929 |
