siRNA Details
Gene Symbol: Rgmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028583.1 | - | 603 | |||
AK030386.1 | - | 3 | 3473 | |||
AK047390.1 | - | 3 | 3473 | |||
BC096024.1 | - | 3 | 3464 | 6821755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
