siRNA Details
Gene Symbol: C130074G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178692.3 | NP_848807.1 | 2 | 854 | ||
GenBank mRNA | AK052654.1 | - | 2 | 818 | ||
AK081755.1 | - | 2 | 833 | |||
AK084264.1 | - | 2 | 827 | |||
AK087640.1 | - | 2 | 845 |
