siRNA Details
Gene Symbol: Slc25a44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145876.2 | NP_001139348.1 | 4 | 3218 | ||
NM_001145877.2 | NP_001139349.1 | 3 | 3136 | |||
NM_001281795.1 | NP_001268724.1 | 4 | 3139 | |||
NM_178696.5 | NP_848811.2 | 4 | 3264 | |||
GenBank mRNA | AK032960.1 | - | 4 | 3242 | ||
AK044648.1 | - | 5 | 3069 | |||
AK046586.1 | - | 4 | 3227 | |||
AK079532.1 | - | 4 | 3228 | |||
AK083273.1 | - | 4 | 3235 | |||
AK083470.1 | - | 4 | 3115 | |||
AK134914.1 | - | 4 | 3264 | |||
AK144731.1 | - | 4 | 3192 | |||
AK165159.1 | - | 3 | 3105 | |||
AK166011.1 | - | 4 | 3236 | |||
AK168955.1 | - | 4 | 3239 | |||
AK170066.1 | - | 1 | 1378 | |||
AK190462.1 | - | 1 | 73 | |||
BC012234.1 | - | 4 | 3061 | 4191504 | ||
BC052771.1 | - | 4 | 3209 | 6310271 | ||
BY639336.1 | - | 1 | 95 |
