siRNA Details
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178709.4 | NP_848824.1 | 14 | 3141 | ||
GenBank mRNA | AK028768.1 | - | 14 | 2776 | ||
AK032376.1 | - | 14 | 2207 | |||
AK033867.1 | - | 13 | 3473 | |||
AK051527.1 | - | 14 | 3142 | |||
AK136229.1 | - | 10 | 1771 | |||
BC064100.1 | - | 4 | 1257 |
