siRNA Details
Gene Symbol: GRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005160.3 | NP_005151.2 | 15 | 1398 | ||
XM_011529975.2 | XP_011528277.1 | 14 | 1108 | |||
GenBank mRNA | AK058103.1 | - | 14 | 1188 | ||
AK292234.1 | - | 15 | 1264 | |||
AK310398.1 | - | 15 | 1266 | |||
BC036797.1 | - | 17 | 1492 | 5590378 | ||
X69117.1 | - | 15 | 1251 |
