siRNA Details
Gene Symbol: ALDH7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001182.4 | NP_001173.2 | 14 | 1490 | ||
NM_001201377.1 | NP_001188306.1 | 14 | 1490 | |||
NM_001202404.1 | NP_001189333.2 | 12 | 1298 | |||
XM_011543417.2 | XP_011541719.1 | 14 | 1481 | |||
XM_017009493.1 | XP_016864982.1 | 14 | 1266 | |||
GenBank mRNA | AK092507.1 | - | 5 | 1502 | ||
AK295526.1 | - | 9 | 725 | |||
AK297365.1 | - | 12 | 1244 | |||
AK312459.1 | - | 14 | 1319 | |||
BC002515.2 | - | 14 | 1307 | 3139235 | ||
BC071712.1 | - | 14 | 1284 | 5518212 | ||
BC073174.1 | - | 14 | 1276 | 6501646 | ||
S74728.1 | - | 14 | 1268 |
