siRNA Details
Gene Symbol: CKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001824.4 | NP_001815.2 | 4, 5 | 648 | ||
XM_017026240.1 | XP_016881729.1 | 3, 4 | 533 | |||
GenBank mRNA | AK129878.1 | - | 4, 5 | 546 | ||
AK300730.1 | - | 4, 5 | 436 | |||
AK313281.1 | - | 4, 5 | 546 | |||
BC007462.1 | - | 4, 5 | 546 | 3934232 | ||
BT006793.1 | - | 3, 4 | 473 | |||
M14780.1 | - | 4, 5 | 551 |
