siRNA Details
Gene Symbol: CLPTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282175.1 | NP_001269104.1 | 14 | 1810 | ||
NM_001282176.1 | NP_001269105.1 | 14 | 1834 | |||
NM_001294.3 | NP_001285.1 | 14 | 1961 | |||
GenBank mRNA | AF037339.1 | - | 14 | 1804 | ||
AK027698.1 | - | 2 | 257 | |||
AK074935.1 | - | 14 | 1805 | |||
AK075262.1 | - | 14 | 1804 | |||
AK223553.1 | - | 14 | 1805 | |||
AK293314.1 | - | 14 | 1826 | |||
AK304480.1 | - | 14 | 1793 | |||
AK316094.1 | - | 14 | 1830 | |||
BC004865.2 | - | 14 | 1840 | 3840991 | ||
BC012359.1 | - | 14 | 1794 | 4685026 | ||
BM676309.1 | - | 1 | 685 | |||
BT007262.1 | - | 14 | 1789 | |||
KU177986.1 | - | 14 | 1789 | |||
KU177987.1 | - | 14 | 1789 |
