siRNA Details
Gene Symbol: EPHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000537.1 | XP_016856026.1 | 1, 2 | 259 | ||
GenBank mRNA | AK296788.1 | - | 1, 2 | 224 | ||
BC037166.2 | - | 1, 2 | 205 | 5090654 | ||
CB991812.1 | - | 1, 2 | 269 | |||
EU826606.1 | - | 1, 2 | 87 | |||
HY005615.1 | - | 1, 2 | 265 | |||
M59371.1 | - | 1, 2 | 200 |
