siRNA Details
Gene Symbol: GPR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005282.2 | NP_005273.1 | 2 | 1494 | ||
XM_017026607.1 | XP_016882096.1 | 2 | 2082 | |||
XM_017026608.1 | XP_016882097.1 | 2 | 1403 | |||
GenBank mRNA | AK290698.1 | - | 2 | 1447 | ||
BC038450.1 | - | 2 | 1482 | |||
BC067535.1 | - | 1 | 853 | 6971751 | ||
BC067536.1 | - | 1 | 853 | 6971752 | ||
EU432116.1 | - | 1 | 549 |
