siRNA Details
Gene Symbol: ITGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002204.3 | NP_002195.1 | 7 | 1572 | ||
GenBank mRNA | AB209658.1 | - | 7 | 1472 | ||
AK289961.1 | - | 7 | 1198 | |||
AK293323.1 | - | 7 | 902 | |||
AK303384.1 | - | 8 | 1126 | |||
BC136636.1 | - | 7 | 1172 | 9020626 | ||
BC144328.1 | - | 7 | 1172 | 9052848 | ||
BC150190.1 | - | 7 | 1415 | 40148521 | ||
D01038.1 | - | 7 | 1004 | |||
M59911.1 | - | 7 | 1173 |
