siRNA Details
Gene Symbol: NOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000620.4 | NP_000611.1 | 2 | 759 | ||
NM_001204218.1 | NP_001191147.1 | 2 | 759 | |||
XM_011538398.2 | XP_011536700.1 | 3 | 630 | |||
XM_017019345.1 | XP_016874834.1 | 2 | 766 | |||
XM_017019346.1 | XP_016874835.1 | 2 | 580 | |||
XM_017019347.1 | XP_016874836.1 | 2 | 669 | |||
GenBank mRNA | AK294435.1 | - | 2 | 563 | ||
AK307481.1 | - | 2 | 759 | |||
D16408.1 | - | 1 | 48 | |||
EU753241.1 | - | 2 | 733 | |||
L02881.1 | - | 1 | 478 | |||
U17327.1 | - | 2 | 733 | |||
U31466.1 | - | 1 | 58 |
