siRNA Details
Gene Symbol: NOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000620.4 | NP_000611.1 | 2 | 900 | ||
NM_001204218.1 | NP_001191147.1 | 2 | 900 | |||
XM_011538398.2 | XP_011536700.1 | 3 | 771 | |||
XM_017019345.1 | XP_016874834.1 | 2 | 907 | |||
XM_017019346.1 | XP_016874835.1 | 2 | 721 | |||
XM_017019347.1 | XP_016874836.1 | 2 | 810 | |||
GenBank mRNA | AK294435.1 | - | 2 | 704 | ||
AK307481.1 | - | 2 | 900 | |||
D16408.1 | - | 1 | 189 | |||
EU753241.1 | - | 2 | 874 | |||
L02881.1 | - | 1 | 619 | |||
U17327.1 | - | 2 | 874 | |||
U31466.1 | - | 1 | 199 |
