siRNA Details
Gene Symbol: RSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012425.3 | NP_036557.1 | 7, 8 | 742 | ||
NM_152724.2 | NP_689937.2 | 6, 7 | 630 | |||
GenBank mRNA | AK292911.1 | - | 6, 7 | 555 | ||
AK312520.1 | - | 7, 8 | 685 | |||
BC005993.1 | - | 7, 8 | 712 | 4050837 | ||
BC008691.1 | - | 6, 7 | 901 | 3872071 | ||
BC015644.2 | - | 7, 8 | 702 | 4647836 | ||
BC107898.1 | - | 6, 7 | 609 | |||
BG400752.1 | - | 6, 7 | 631 | |||
BX647733.1 | - | 6, 7 | 606 | |||
CR536521.1 | - | 6, 7 | 599 | |||
CR541840.1 | - | 6, 7 | 599 |
