siRNA Details
Gene Symbol: CAPN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005632.2 | NP_005623.1 | 4, 5 | 1819 | ||
XM_011522620.2 | XP_011520922.1 | 5, 6 | 5118 | |||
XM_011522621.2 | XP_011520923.1 | 5, 6 | 6046 | |||
XM_011522622.1 | XP_011520924.1 | 3, 4 | 4750 | |||
XM_011522623.2 | XP_011520925.1 | 6, 7 | 2821 | |||
XM_011522624.2 | XP_011520926.1 | 5, 6 | 2159 | |||
XM_011522625.2 | XP_011520927.1 | 4, 5 | 2045 | |||
XM_011522626.2 | XP_011520928.1 | 5, 6 | 5610 | |||
XM_011522627.2 | XP_011520929.1 | 5, 6 | 5615 | |||
XM_011522628.2 | XP_011520930.1 | 5, 6 | 2679 | |||
XM_011522629.2 | XP_011520931.1 | 5, 6 | 2542 | |||
XM_011522630.2 | XP_011520932.1 | 4, 5 | 1880 | |||
XM_011522631.2 | XP_011520933.1 | 3, 4 | 1766 | |||
XM_011522632.2 | XP_011520934.1 | 3, 4 | 2197 | |||
XM_017023596.1 | XP_016879085.1 | 6, 7 | 2958 | |||
GenBank mRNA | U85647.1 | - | 4, 5 | 1819 |
