siRNA Details
Gene Symbol: SSR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204526.1 | NP_001191455.1 | 3 | 257 | ||
NM_001204527.1 | NP_001191456.1 | 3 | 280 | |||
NM_006280.2 | NP_006271.1 | 2 | 230 | |||
NR_037927.1 | 4 | 451 | ||||
XM_011531186.1 | XP_011529488.1 | 3 | 139 | |||
XM_011531187.1 | XP_011529489.1 | 3 | 139 | |||
XM_017029756.1 | XP_016885245.1 | 3 | 1028 | |||
XM_017029757.1 | XP_016885246.1 | 3 | 763 | |||
GenBank mRNA | AK057117.1 | - | 1 | 1813 | ||
AK094395.1 | - | 3 | 1028 | |||
AK290493.1 | - | 2 | 136 | |||
BC003371.1 | - | 2 | 110 | 2900758 | ||
BE791319.1 | - | 2 | 152 | |||
BG165852.1 | - | 3 | 264 | |||
BP202705.1 | - | 3 | 219 | |||
BT007192.1 | - | 2 | 106 | |||
CD672615.1 | - | 3 | 322 | |||
DB238237.1 | - | 2 | 230 | |||
X90583.1 | - | 2 | 136 | |||
Z69043.1 | - | 2 | 155 |
